Phosphoryl trichloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phosphorus (V) oxide chloride, Phosphorus oxychloride, Phosphoryl chloride, Phosphorus(V) oxychloride

19129-0010
TS19129-0010EA 130 USD
TS19129-0010
Phosphoryl trichloride 99%
Phosphoryl trichloride
Formula: POCl₃
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: 1.2 °C
Density: 1.645 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011443
CAS Number: 10025-87-3
EINECS: 233-046-7
UN: 1810
ADR: 6.1,I
Merck Index: 12,07506

Order Now

Specification Test Results

Appearance Clear liquid
Refractive index 1.4600 to 1.4620 (20°C, 589 nm)
Titration Argentometric ≥98.5 % (ex Cl)
Color scale ≤40 APHA
Phosphorus trichloride (PCl3) ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR