Sodium borohydride 0.5 M in diglyme, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

191131000
TS191131000EA 82 USD
TS191131000
Sodium borohydride 0.5 M in diglyme, AcroSeal®
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400
Density: 0.945 g/cm³
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
Merck Index: 13,08664

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.45 to 0.55 M
Solution contains 18.915 g NaBH4/l C6H14O3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR