Aluminium oxide neutral Brockmann Grade I 50-200 µm, 90A for chromatography

Supplier: Thermo Scientific Chemicals

AC19041-0010
TS19041-0010EA 110 USD
TS19041-0010
Aluminium oxide neutral Brockmann Grade I 50-200 µm, 90A for chromatography
Aluminium oxide neutral
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C (760 mmHg)
Melting Pt: 2050 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Particle size ≤5 % (> 200 um)
Specific surface area 90 to 170 m2/g,N2 adsorp
pH 6.5 to 7.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR