Chromeazurol S (Mordant blue 29)

Supplier: Thermo Scientific Chemicals
Warning

19005-1000 19005-0250
TS19005-1000EA 289 USD
TS19005-1000 TS19005-0250
Chromeazurol S (Mordant blue 29)
Chromeazurol S (Mordant blue 29)
Formula: C₂₃H₁₆Cl₂O₉S
Storage Temperature: Ambient
MDL Number: MFCD00001615
CAS Number: 1667-99-8

Order Now

Specification Test Results

Appearance Dark brown to black Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
UV Lambda max 424 to 436 nm (c=0.03 g/l in H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR