Bromotrimethylsilane 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylbromosilane, Trimethylsilyl bromide

AC18665-0050 AC18665-0250 AC18665-1000 AC18665-5000
TS18665-0050EA 55 USD
TS18665-0050 TS18665-0250 TS18665-1000 TS18665-5000
Bromotrimethylsilane 98%
Bromotrimethylsilane
Formula: C₃H₉BrSi
MW: 153.09 g/mol
Boiling Pt: 79
Melting Pt: -43
Density: 1.16 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00000048
CAS Number: 2857-97-8
EINECS: 220-672-0
UN: 2920
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4250 (20°C, 589 nm)
Titration Argentometric ≥97.5 %
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR