1,3-Dibromobenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
18664-0100
18664-0500
TS18664-0100EA
36
USD
TS18664-0100
TS18664-0500
1,3-Dibromobenzene 97%
1,3-Dibromobenzene
Formula:
C₆H₄Br₂ Boiling Pt: 218…219 °C (760 mmHg) Melting Pt: –7 °C Density: 1.95 g/cm³ Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000078 CAS Number: 108-36-1 EINECS: 203-574-2 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6070 to 1.6090 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...