1,3-Dibromobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

18664-0100 18664-0500
TS18664-0100EA 36 USD
TS18664-0100 TS18664-0500
1,3-Dibromobenzene 97%
1,3-Dibromobenzene
Formula: C₆H₄Br₂
Boiling Pt: 218…219 °C (760 mmHg)
Melting Pt: –7 °C
Density: 1.95 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000078
CAS Number: 108-36-1
EINECS: 203-574-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6070 to 1.6090 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR