Methanol-[D₄] 100.0 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

186450100
TS186450100EA 344 USD
TS186450100
Methanol-[D₄] 100.0 atom % D for NMR spectroscopy
Methanol-[D4]
Formula: D₃COD
MDL Number: MFCD00044637
CAS Number: 811-98-3
UN: 1230

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR