Borane dimethyl sulphide complex 1 mol/l in dichloromethane, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Borane-methyl sulfide complex, Borane-methyl sulphide complex

18561-1000
TS18561-1000EA 94 USD
TS18561-1000
Borane dimethyl sulphide complex 1 mol/l in dichloromethane, AcroSeal™
Borane dimethyl sulphide complex
Formula: C₂H₉BS
MW: 75.97 g/mol
Flash Pt: 18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013189
CAS Number: 13292-87-0
EINECS: 236-313-6
UN: 3399

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.95 to 1.05 M
Water ≤3 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR