Deuterium oxide 100.0 atom % D, AcroSeal™ for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

Synonyms: Water-D2, Heavy water

18476-1000
TS18476-1000EA 396 USD
TS18476-1000
Deuterium oxide 100.0 atom % D, AcroSeal™ for NMR spectroscopy
Deuterium oxide
Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101.4 °C (760 mmHg)
Melting Pt: 3.8 °C
Density: 1.105 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00044636
CAS Number: 7789-20-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
D-Enrichment ≥99.95 %
Melting point 143 to 152 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %
Titration with NaOH 18.5 to 19.5 % (Fumaric acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR