tert-Butylchlorodimethylsilane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride
18393-1000
18393-5000
AC18393-0050
AC18393-0250
TS18393-1000EA
307
USD
TS18393-1000
TS18393-5000
TS18393-0050
TS18393-0250
tert-Butylchlorodimethylsilane 98%
tert-Butylchlorodimethylsilane
Formula:
C₆H₁₅ClSi Boiling Pt: 125 °C (1013 hPa) Melting Pt: 85…91 °C Density: 0.87 g/cm³ Flash Pt: 22 °C |
MDL Number:
MFCD00000501 CAS Number: 18162-48-6 EINECS: 242-042-4 UN: 2921 |
Specification Test Results
Appearance | Colorless to white Crystals |
Infrared spectrum | Conforms |
Melting point | 85 to 91 °C |
GC | ≥97.5 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...