Polyethylene glycol, M.W. 600,000 - 1,000,000
Supplier: Thermo Scientific Chemicals
183220050
183221000
183225000
TS183220050EA
29
USD
TS183220050
TS183221000
TS183225000
Polyethylene glycol, M.W. 600,000 - 1,000,000
Polyethylene glycol
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (10 g, 5 h, 10 mmHg) |
Viscosity | 300 to 600 mPa.s (2 % w/w in water at 25 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...