Polyethylene glycol, M.W. 600,000 - 1,000,000

Supplier: Thermo Scientific Chemicals

183220050 183221000 183225000
TS183220050EA 29 USD
TS183220050 TS183221000 TS183225000
Polyethylene glycol, M.W. 600,000 - 1,000,000
Formula: H(OCH₂CH₂)nOH
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (10 g, 5 h, 10 mmHg)
Viscosity 300 to 600 mPa.s (2 % w/w in water at 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR