Polyvinyl alcohol, M.W. ∼ 31,000-50,000 85-89 %, hydrolysed

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

183140010 183140025 183141000
TS183140010EA 125 USD
TS183140010 TS183140025 TS183141000
Polyvinyl alcohol, M.W. ∼ 31,000-50,000 85-89 %, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Melting Pt: 230…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Appearance White to cream Granular powder
Infrared spectrum Conforms
Viscosity 5.2 to 6.2 cP (4 % in water)
Degree of hydrolysis 87.0 to 89.0 mol %
Ash ≤0.7 % (as Na2O)
pH 4.5 to 6.5 (4 % in water)
Volatile matter ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR