N,N-Diethylformamide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Formic acid diethylamide, Formdiethylamide

181375000 181370250 181371000
TS181375000EA 412 USD
TS181375000 TS181370250 TS181371000
N,N-Diethylformamide 99%
N,N-Diethylformamide
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 176
Density: 0.9 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00003287
CAS Number: 617-84-5
EINECS: 210-533-2
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥92.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR