Formamide 99.5+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Carbamaldehyde

181090010 181090025 181090250
TS181090010EA 129 USD
TS181090010 TS181090025 TS181090250
Formamide 99.5+%, extra pure
Formamide
Formula: HCONH₂
Boiling Pt: 210 °C (760 mmHg)
Melting Pt: 2…3 °C
Density: 1.133 g/cm³
Flash Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4480 (20°C, 589 nm)
Water ≤0.1 % (Coulometric)
GC ≥99.50 %
Residue after evaporation ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR