1-Chloronaphthalene 85% remainder 2-Chloronaphthalene, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

18097-0010 18097-0025 18097-1000
TS18097-0010EA 318 USD
TS18097-0010 TS18097-0025 TS18097-1000
1-Chloronaphthalene 85% remainder 2-Chloronaphthalene, Technical Grade
1-Chloronaphthalene
Formula: C₁₀H₇Cl
Boiling Pt: 259 °C (760 mmHg)
Melting Pt: –2.5 °C
Density: 1.194 g/cm³
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00003874
CAS Number: 90-13-1
EINECS: 201-967-3
UN: 3082
ADR: 9,III
Merck Index: 13,02168

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.6300 to 1.6340 (20°C, 589 nm)
HPLC ≥93 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR