Ethylene glycol monomethyl ether (2-methoxyethanol) 99+%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethylene glycol monomethyl ether
18079-0025
18079-0010
180790250
TS18079-0025EA
128
USD
TS18079-0025
TS18079-0010
TS180790250
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+%, extra pure
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula:
C₃H₈O₂ Boiling Pt: 124.6 °C (1013 hPa) Melting Pt: –85 °C Density: 0.96 g/cm³ Flash Pt: 38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002867 CAS Number: 109-86-4 EINECS: 203-713-7 UN: 1188 ADR: 3,III Merck Index: 12,06118 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4010 to 1.4040 (20 °C, 589 nm) |
Water | ≤0.2 % |
GC | ≥99.0 % |
Color scale | ≤15 APHA |
Residue after evaporation | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...