Polyethylene glycol, M.W. 250,000 - 400,000

Supplier: Thermo Scientific Chemicals

17860-0050 178600010 178602500
TS17860-0050EA 24 USD
TS17860-0050 TS178600010 TS178602500
Polyethylene glycol, M.W. 250,000 - 400,000
Polyethylene glycol
Formula: H(OCH₂CH₂)nOH
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR