Polyethylene glycol, M.W. 200,000 - 300,000
Supplier: Thermo Scientific Chemicals
17859-0010
178590050
178592500
TS17859-0010EA
907
USD
TS17859-0010
TS178590050
TS178592500
Polyethylene glycol, M.W. 200,000 - 300,000
Polyethylene glycol
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (10 g, 50°C, 5 h, 10 mmHg) |
Viscosity | 400 to 800 mPa.s (10 % w/w in water at 25°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...