Polyethylene oxide, M.W. 100000-200000
Supplier: Thermo Scientific Chemicals
Synonyms:
PEO
178580050
178582500
178585000
TS178580050EA
27
USD
TS178580050
TS178582500
TS178585000
Polyethylene oxide, M.W. 100000-200000
Polyethylene oxide
Formula:
(-CH₂CH₂O-)n Melting Pt: 65…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00147970 CAS Number: 25322-68-3 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (10 g, 50 °C, 5 h, 10 mmHg) |
Viscosity | 80 to 200 mPa.s (10 % w/w in water at 25 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...