Hexane (mixture of isomers) 98+%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
176660010
176660025
176660250
TS176660010EA
55
USD
TS176660010
TS176660025
TS176660250
Hexane (mixture of isomers) 98+%, extra pure
Hexane (mixture of isomers)
Formula:
H₃C(CH₂)₄CH₃ Boiling Pt: 69 °C (1013 hPa) Melting Pt: –95 °C Density: 0.659 g/cm³ Flash Pt: –22 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179311 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3780 to 1.3820 (20°C, 589 nm) |
Water | ≤0.05 % |
GC | ≥98.0 % (sum of isomers) |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Free acid (CH3COOH) | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...