Tetrahydrofuran 99+% stabilized, extra pure

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

176630010 176630025 176630100 176630250
TS176630010EA 97 USD
TS176630010 TS176630025 TS176630100 TS176630250
Tetrahydrofuran 99+% stabilized, extra pure
Tetrahydrofuran
Formula: C₄H₈O
Boiling Pt: 66 °C (760 mmHg)
Melting Pt: –108.4 °C
Density: 0.88 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer 200 to 400 ppm BHT
Residue after evaporation ≤0.005 %
Peroxides ≤0.0025 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR