Tetraethylammonium hydroxide 20% (w/w) in water

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetrylammonium hydroxide

176072500 176070010 176070050
TS176072500EA 97 USD
TS176072500 TS176070010 TS176070050
Tetraethylammonium hydroxide 20% (w/w) in water
Tetraethylammonium hydroxide
Formula: C₈H₂₁NO
MW: 147.26
Density: 1.02 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00009024
CAS Number: 77-98-5
EINECS: 201-073-3
UN: 2922
ADR: 8 II
Merck Index: 12,09342

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Titration with HCl ≥20 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR