Deuterium chloride 20% (w/w) in D₂O 99+ atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

17542-0500
TS17542-0500EA 156 USD
TS17542-0500
Deuterium chloride 20% (w/w) in D₂O 99+ atom % D for NMR spectroscopy
Deuterium chloride
Formula: DCl
Density: 1.2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00044642
CAS Number: 7698-05-7
EINECS: 231-715-8
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless Solution
D-Enrichment ≥99.0 %
Concentration 20 to 22 % w/w
Ash ≤0.3 % (1000°C, 0.5 h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR