Silver trifluoromethanesulfonate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Silver trifluoromethanesulfonate, Trifluoromethanesulphonic acid silver salt, Trifluoromethanesulfonic acid silver salt

AC17520-0100 AC17520-0250 AC17520-1000
TS17520-0100EA 146 USD
TS17520-0100 TS17520-0250 TS17520-1000
Silver trifluoromethanesulfonate 99+%
Silver trifluoromethanesulphonate
Formula: CAgF₃O₃S
MW: 256.94 g/mol
Melting Pt: 356
Storage Temperature: Ambient
MDL Number: MFCD00013226
CAS Number: 2923-28-6
EINECS: 220-882-2

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Elemental Analysis ≥99.0 %
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR