4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98% free radical

Supplier: Thermo Scientific Chemicals
Danger

17505-0010 17505-0050 17505-0250
TS17505-0010EA 33 USD
TS17505-0010 TS17505-0050 TS17505-0250
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98% free radical
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-Hydroxy-TEMPO, TEMPOL, free radical)
Formula: C₉H₁₈NO₂
Melting Pt: 69…73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006478
CAS Number: 2226-96-2
EINECS: 218-760-9

Order Now

Specification Test Results

Appearance Orange Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 69 to 73 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR