4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98% free radical
Supplier: Thermo Scientific Chemicals
|
Danger
|
17505-0010
17505-0050
17505-0250
TS17505-0010EA
33
USD
TS17505-0010
TS17505-0050
TS17505-0250
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98% free radical
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-Hydroxy-TEMPO, TEMPOL, free radical)
Formula:
C₉H₁₈NO₂ Melting Pt: 69…73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006478 CAS Number: 2226-96-2 EINECS: 218-760-9 |
Specification Test Results
Appearance | Orange Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 69 to 73 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...