Mercury(II) acetate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt

AC17502-0025 AC17502-5000
TS17502-0025EA 1640 USD
TS17502-0025 TS17502-0050 TS17502-5000
Mercury(II) acetate 98%
Mercury (II) acetate
Formula: (H₃CCOO)₂Hg
Melting Pt: 179…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 179 to 182 °C
Titration Complexometric ≥97.5 %
Chloride (Cl) ≤300 ppm
Sulfate (SO4) ≤100 ppm
Iron (Fe) ≤50 ppm
Mercurous Mercury (as Hg) ≤8000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR