2,5-Dimethylpyrazine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17452-0050
17452-0250
TS17452-0050EA
39
USD
TS17452-0050
TS17452-0250
2,5-Dimethylpyrazine 99%
2,5-Dimethylpyrazine
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 155 Melting Pt: 15 Density: 0.99 g/cm³ Flash Pt: 63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006147 CAS Number: 123-32-0 EINECS: 204-618-3 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4987 to 1.5007 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...