2,5-Dimethylpyrazine 99%

Supplier: Thermo Scientific Chemicals
Warning

17452-0050 17452-0250
TS17452-0050EA 39 USD
TS17452-0050 TS17452-0250
2,5-Dimethylpyrazine 99%
2,5-Dimethylpyrazine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 155
Melting Pt: 15
Density: 0.99 g/cm³
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00006147
CAS Number: 123-32-0
EINECS: 204-618-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4987 to 1.5007 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR