3-Aminobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Danger

17304-0250 17304-1000 17304-0050
TS17304-0250EA 124 USD
TS17304-0250 TS17304-1000 TS17304-0050
3-Aminobenzonitrile 99%
3-Aminobenzonitrile
Formula: C₇H₆N₂
Boiling Pt: 288…290 °C (1013 hPa)
Melting Pt: 50…54 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007756
CAS Number: 2237-30-1
EINECS: 218-800-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown Crystalline solid
Infrared spectrum Conforms
Melting point 50 to 54 °C
GC ≥98.5 %
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR