Tritylium tetrafluoroborate 97%

Supplier: Thermo Scientific Chemicals
Danger

17295-0050 17295-0250 17295-1000
TS17295-0050EA 45 USD
TS17295-0050 TS17295-0250 TS17295-1000
Tritylium tetrafluoroborate 97%
Tritylium tetrafluoroborate
Formula: C₁₉H₁₅BF₄
Melting Pt: 205…215 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00013120
CAS Number: 341-02-6
EINECS: 206-433-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to green to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 203 to 213 °C (Dec)
NMR ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR