Bicine 99+% for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

17265-0010 17265-1000 17265-2500
TS17265-0010EA 525 USD
TS17265-0010 TS17265-1000 TS17265-2500
Bicine 99+% for analysis
Bicine
Formula: C₆H₁₃NO₄
Melting Pt: 190…192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS Number: 150-25-4
EINECS: 205-755-1

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or needles
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
Sulfated ash ≤0.1 %
UV at 250 nm A: ≤0.05
pH 4.0 to 5.5 (10 % at 20°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR