γ-(N-Morpholino)propanesulfonic acid (MOPS) 99% for biochemistry

Supplier: Thermo Scientific Chemicals

17263-0025 17263-0250 17263-1000 17263-5000
TS17263-0025EA 1374 USD
TS17263-0025 TS17263-0250 TS17263-1000 TS17263-5000
γ-(N-Morpholino)propanesulfonic acid (MOPS) 99% for biochemistry
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula: C₇H₁₅NO₄S
Melting Pt: 277…282 °C
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1.0 %
UV at 280 nm A: ≤0.03
pH 3.0 to 4.5 (1 % in water at 25°C)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR