(+)-Dimethyl-L-tartrate 99%

Supplier: Thermo Scientific Chemicals

17249-0250 17249-1000 17249-5000
TS17249-0250EA 32 USD
TS17249-0250 TS17249-1000 TS17249-5000
(+)-Dimethyl-L-tartrate 99%
(+)-Dimethyl L-tartrate
Formula: C₆H₁₀O₆
Boiling Pt: 157…159 °C (15hPa)
Melting Pt: 48…50 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00064437
CAS Number: 608-68-4
EINECS: 210-166-8

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation +20° to +22° (20°C, 589 nm) (10% in water)
Elemental Analysis 5.22 to 5.65 % (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR