4-Chloro-7-nitrobenzofurazan 98%

Supplier: Thermo Scientific Chemicals
Warning

17239-0010 17239-0050 17239-0250
TS17239-0010EA 33 USD
TS17239-0010 TS17239-0050 TS17239-0250
4-Chloro-7-nitrobenzofurazan 98%
4-Chloro-7-nitrobenzofurazan
Formula: C₆H₂ClN₃O₃
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00005808
CAS Number: 10199-89-0
EINECS: 233-496-4

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 99 °C
Sulfated ash ≤0.1 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR