Dimethyl glyoxime 99+%

Supplier: Thermo Scientific Chemicals
Danger

17200-0025 17200-1000 17200-5000
TS17200-0025EA 494 USD
TS17200-0025 TS17200-1000 TS17200-5000
Dimethyl glyoxime 99+%
Dimethyl glyoxime
Formula: CH₃C(=NOH)C(=NOH)CH₃
Melting Pt: 239…242 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
EINECS: 202-420-1
UN: 2926
ADR: 4.1,III
Merck Index: 1303272

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 239 to 242 °C (Dec)
Sulfated ash ≤0.1 %
Gravimetry ≥99.0 %
Insoluble matter ≤0.05 % (in alcohol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR