Charcoal activated, NORIT® SA 2, decolorizing

Supplier: Thermo Scientific Chemicals

17153-0010 171530050
TS17153-0010EA 114 USD
TS17153-0010 TS171530050
Charcoal activated, NORIT® SA 2, decolorizing
Charcoal activated
Formula: C
Melting Pt: 3652 °C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specification Test Results

Appearance Black Powder
Loss on drying ≤10 %
Molasses number ≤390
Water ≤1.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR