EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) 98+%

Supplier: Thermo Scientific Chemicals
Danger

17144-1000 17144-0010 17144-0100 17144-0500 17144-5000
TS17144-1000EA 901 USD
TS17144-1000 TS17144-0010 TS17144-0100 TS17144-0500 TS17144-5000
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) 98+%
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride
Formula: C₈H₁₇N₃·HCl
Melting Pt: ∼ 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012503
CAS Number: 25952-53-8
EINECS: 247-361-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 116 °C
Titration Argentometric ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR