3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(2-Pyridyl)-5,6-bis(4-sulfophenyl)-1,2,4-triazine disodium salt hydrate, 3-(2-Pyridyl)-5,6-bis(4-sulphophenyl)-1,2,4-triazine disodium salt hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulphonic acid disodium salt hydrate, PDT disulphonate hydrate, FerroZine(TM) Iron reagent disodium salt hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate, PDT disulfonate hydrate

17101-0010 17101-0050 17101-0250
TS17101-0010EA 56 USD
TS17101-0010 TS17101-0050 TS17101-0250
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate 97+%
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate
Formula: C₂₀H₁₂N₄Na₂O₆S₂·xH₂O
MW: 514.44 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00717621
CAS Number: 28048-33-1
EINECS: 248-797-6

Order Now

Specification Test Results

Appearance Yellow to yellow-green Powder
Infrared spectrum Conforms
Water ≤10.0 % (K.F.)
Sulfated ash 20.0 % to 27.8 %
Solubility (5 % acetate-buffer pH 5.5) Clear yellow
UV Lambda max 280 to 288 nm (in water)
Specific extinction E (1%/1cm) 385 to 465 (at lambda max)
Titration with HClO4 ≥97.0 % (On dried substance)
Suitability (for Cu + Fe determination) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR