1-(4-Chlorophenyl)cyclopropanecarboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

17069-0100 17069-0500
TS17069-0100EA 119 USD
TS17069-0100 TS17069-0500
1-(4-Chlorophenyl)cyclopropanecarboxylic acid 99%
1-(4-Chlorophenyl)cyclopropanecarboxylic acid
Formula: C₁₀H₉ClO₂
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00001289
CAS Number: 72934-37-3
EINECS: 277-105-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 152 to 157 °C
Titration with NaOH ≥98.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR