1,3-Dithiane 97%

Supplier: Thermo Scientific Chemicals

16951-0050 16951-0250 16951-1000
TS16951-0050EA 51 USD
TS16951-0050 TS16951-0250 TS16951-1000
1,3-Dithiane 97%
1,3-Dithiane
Formula: C₄H₈S₂
Melting Pt: 49…54 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00006654
CAS Number: 505-23-7
EINECS: 208-006-7

Order Now

Specification Test Results

Appearance White to light beige Powder or crystals or needles
Infrared spectrum Conforms
Melting point 49 to 54 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR