Dimethyl sulfoxide-[D₆] 100.0 atom % D, > 99.96 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

169020250
TS169020250EA 216 USD
TS169020250
Dimethyl sulfoxide-[D₆] 100.0 atom % D, > 99.96 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
Boiling Pt: 189 °C (5 mmHg)
Melting Pt: 20.2 °C
Density: 1.19 g/cm³
Flash Pt: 88 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR