Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethylene glycol monomethyl ether

168580010 168580025
TS168580010EA 216 USD
TS168580010 TS168580025
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula: C₃H₈O₂
Boiling Pt: 124.6 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.96 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 %
GC ≥99.0 %
UV at 280 nm A: ≤0.05
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity ≤0.0008 meq/g
Peroxides ≤0.002 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR