Methacrylic acid 99.5% stabilised, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylpropenoic acid
16831-0040
16831-0251
16831-0250
168310010
TS16831-0040EA
163
USD
TS16831-0040
TS16831-0251
TS16831-0010
TS16831-0250
TS16831-2500
TS16831-5000
TS168310010
Methacrylic acid 99.5% stabilised, extra pure
Methacrylic acid
Formula:
C₄H₆O₂ MW: 86.09 g/mol Boiling Pt: 163 Melting Pt: 15 Density: 1.01 g/cm³ Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002651 CAS Number: 79-41-4 EINECS: 201-204-4 UN: 2531 ADR: 8 II Merck Index: 13,05967 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4295 to 1.4315 (20°C, 589 nm) |
Water | ≤0.2 % |
GC | ≥99.4 % |
Color scale | ≤20 APHA |
Stabilizer | ca. 250 ppm MEHQ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...