Nitromethane 96% for spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

16786-0025 16786-5000
TS16786-0025EA 336 USD
TS16786-0025 TS16786-5000
Nitromethane 96% for spectroscopy
Nitromethane
Formula: CH₃NO₂
Boiling Pt: 101.2 °C (1013 hPa)
Melting Pt: –29 °C
Density: 1.12 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00007400
CAS Number: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: 3,II
Merck Index: 13,06644

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.03 %
GC ≥95 %
UV at 430 nm A: ≤0.01
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR