N,N-Dimethylformamide 99.8% for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

167790010 167790025 167795000
TS167790010EA 150 USD
TS167790010 TS167790025 TS167795000
N,N-Dimethylformamide 99.8% for spectroscopy
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.945 g/cm³
Flash Pt: 58 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
GC ≥99.75 %
UV 1 cm cell vs water
Color scale ≤15 APHA
Residue after evaporation ≤0.0015 %
Acidity ≤0.001 meq/g
Alkalinity ≤0.001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR