Isobutanol 99+%, Spectrophotometric Grade

Supplier: Thermo Scientific Chemicals
Danger

167700010 167705000
TS167700010EA 168 USD
TS167700010 TS167705000
Isobutanol 99+%, Spectrophotometric Grade
Isobutanol
Formula: (CH₃)₂CHCH₂OH
Boiling Pt: 107.9 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0.8 g/cm³
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 %
GC ≥99.0 %
UV at 330 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR