Acetonitrile 99+% for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

167650010 167650025 167652500
TS167650010EA 184 USD
TS167650010 TS167650025 TS167652500
Acetonitrile 99+% for spectroscopy
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81…82 °C (760 mmHg)
Melting Pt: –46 °C
Density: 0.781 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.03 %
GC ≥99.0 %
UV at 250 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR